Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrafluoro-1-propanol ≥98.0%

Supplier: Decon Laboratories Ltd

T0101-25G T0101-500G T0101-100G
TCT0101-25GEA 36.6 USD InStock
TCT0101-25G TCT0101-500G TCT0101-100G
2,2,3,3-Tetrafluoro-1-propanol ≥98.0%
2,2,3,3-Tetrafluoro-1-propanol
CAS Number: 76-37-9
MDL Number: MFCD00004676
Molecular Formula: C3H4F4O
Molecular Weight: 132.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 106
Flash Point (°C): 45
Specific Gravity (20/20): 1.49


Formula: C₃H₄F₄O
MW: 132.06 g/mol
Boiling Pt: 109
Melting Pt: -15
Density: 1.49 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00004676
CAS Number: 76-37-9
UN: 1987
ADR: 3 III
Merck Index: 12,09346
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dc51da2ec27680ef867d2f805c6608627dc32e8&vid=8dc51da2ec27680ef867d2f805c6608627dc32e8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty