Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoroacetophenone ≥98.0%

Supplier: Decon Laboratories Ltd

T0848-25G T0848-5G
TCT0848-25GEA 139.35 USD InStock
TCT0848-25G TCT0848-5G
α,α,α-Trifluoroacetophenone ≥98.0%
α,α,α-Trifluoroacetophenone
CAS Number: 434-45-7
MDL Number: MFCD00000420
Molecular Formula: C8H5F3O
Molecular Weight: 174.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 166
Flash Point (°C): 41
Specific Gravity (20/20): 1.28


Formula: C₈H₅F₃O
Boiling Pt: 166 °C
Density: 1.28 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00000420
CAS Number: 434-45-7
UN: 1224
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e57914d51cd97795b56ff0a06e00d4e204f7165&vid=1e57914d51cd97795b56ff0a06e00d4e204f7165&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty