Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triphenylantimony oxide ≥95.0% (by HPLC)

Supplier: Decon Laboratories Ltd

T1850-1G T1850-5G
TCT1850-1GEA 152.38 USD InStock
TCT1850-1G TCT1850-5G
Triphenylantimony oxide ≥95.0% (by HPLC)
Triphenylantimony oxide
CAS Number: 4756-75-6
MDL Number: MFCD00013837
Molecular Formula: C6H5OSb
Molecular Weight: 369.08
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Storage Temperature: 0-10°C

Other Product Names / Synonyms: Triphenylstibine Oxide


Formula: C₁₈H₁₅OSb
MDL Number: MFCD00013837
CAS Number: 4756-75-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbecb0711dd13bd4f7744047efa0f1e2da78f34c&vid=dbecb0711dd13bd4f7744047efa0f1e2da78f34c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty