Triisopropylsilane 98
Supplier: BEANTOWN CHEMICAL - BTC
121575-100G
121575-25G
121575-5G
BT121575-100GEA
313.64
USD
BT121575-100G
BT121575-25G
BT121575-5G
Triisopropylsilane 98
Triisopropylsilane
CAS: 6485-79-6; MDL No: MFCD00009657
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: [(CH3)2CH]3SiH; Molecular Formula: C9H22Si; MW: 158.36
Boiling Point: 166°; Flash point: 35°C (95°F)
Density (g/mL): 0.773; Refractive Index: 1.434
Moisture Sensitive
- Refractive index: 1.434
- Moisture Sensitive
- Liquid
Formula:
C₉H₂₂Si MW: 158.36 g/mol Boiling Pt: 84 Density: 0.773 g/mL Flash Pt: 35 °C (95 °F) |
MDL Number:
MFCD00009657 CAS Number: 6485-79-6 ADR: 3 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...