Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carbethoxyhexafluorobutyryl chloride ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

121605-1G 121605-250MG 121605-5G
BT121605-1GEA 99 USD InStock
BT121605-1G BT121605-250MG BT121605-5G
4-Carbethoxyhexafluorobutyryl chloride ≥97%
4-Carbethoxyhexafluorobutyryl chloride
CAS: 18381-53-8; MDL No: MFCD00054671 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H5ClF6O3; MW: 286.56 Boiling Point: 157-160° Density (g/mL): 1.47 Moisture Sensitive

  • Moisture Sensitive
  • Liquid


Formula: C₇H₅ClF₆O₃
Boiling Pt: 157…160 °C
Density: 1.47 g/mL
MDL Number: MFCD00054671
CAS Number: 18381-53-8
ADR: 8,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty