Methanol absolute ≥99.8% ACS low acid
Supplier: BEANTOWN CHEMICAL - BTC
|
Danger
|
CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000
UN No: UN1230; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3OH; MW: 32.04
Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F)
Density (g/mL): 0.791; Refractive Index: 1.329
- RTECS: PC1400000
- Refractive index: 1.329
- Liquid
Formula:
CH₃OH MW: 32.04 g/mol Boiling Pt: 64.6 Melting Pt: -98 Density: 0.791 g/mL Flash Pt: 9.7 °C (49.5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3 II REACH: 01-2119433307-44 Merck Index: 13,05984 |