Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)phenyl isocyanate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

124765-1G 124765-25G 124765-5G
BT124765-1GEA 54 USD InStock
BT124765-1G BT124765-25G BT124765-5G
4-(Trifluoromethoxy)phenyl isocyanate ≥97%
4-(Trifluoromethoxy)phenyl isocyanate
CAS: 35037-73-1; EC No: 252-328-0; MDL No: MFCD00035702 UN No: UN2206; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C8H4F3NO2; MW: 203.12 Boiling Point: 78°/22 mmHg; Flash point: 69°C (156°F) Density (g/mL): 1.346; Refractive Index: 1.458 Moisture Sensitive

  • Refractive index: 1.458
  • Moisture Sensitive
  • Liquid


Formula: C₈H₄F₃NO₂
Boiling Pt: 78 °C/22 mmHg
Density: 1.346 g/mL
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00035702
CAS Number: 35037-73-1
EINECS: 252-328-0
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty