Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexachloroplatinic(IV) acid hexahydrate ≥99.95% ACS

Supplier: BEANTOWN CHEMICAL - BTC

128610-1G 128610-25G 128610-5G
BT128610-1GEA 148.36 USD InStock
BT128610-1G BT128610-25G BT128610-5G
Hexachloroplatinic(IV) acid hexahydrate ≥99.95% ACS
Hexachloroplatinic(IV) acid hexahydrate
CAS: 18497-13-7; EC No: 241-010-7; MDL No: MFCD00149910; RTECS: TP1500000 UN No: UN2507; Haz Class: 8; Packing Group: III Lump; Linear Formula: H2PtCl6·6H2O; MW: 517.91 Melting Point: 60° (decomposes) Hygroscopic

  • RTECS: TP1500000
  • Hygroscopic
  • Lump


Formula: H₂PtCl₆·6H₂O
Melting Pt: 60 °C (dec.)
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Order Now
Specification Test Results
Assay 99.95%
Assay, Pt ≥37.50%
Solubility in alcohol Passes test
Alkali and other salts ≤0.05%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty