129610-1L
129610-4L
129610-4X4L
BT129610-1LEA
39
USD
InStock
BT129610-1L
BT129610-4L
BT129610-4X4L
Methanol ≥99.8% for spectrophotometry
Methanol
CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000
UN No: UN1230; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3OH; MW: 32.04
Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F)
Density (g/mL): 0.791; Refractive Index: 1.329
- RTECS: PC1400000
- Refractive index: 1.329
- Liquid