Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Mercaptobenzothiazole ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

129955-100G 129955-500G
BT129955-100GEA 27 USD InStock
BT129955-100G BT129955-500G
2-Mercaptobenzothiazole ≥97%
2-Mercaptobenzothiazole
CAS: 149-30-4; EC No: 205-736-8; MDL No: MFCD00005781; RTECS: DL6475000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Molecular Formula: C7H5NS2; MW: 167.25 Melting Point: 177-181°; Flash point: 200°C (392°F) Air Sensitive

  • RTECS: DL6475000
  • Air Sensitive
  • Crystalline


Formula: C₇H₅NS₂
Melting Pt: 177…181 °C
Flash Pt: 200 °C (392 °F)
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty