Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzotrifluoride ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

130460-1KG 130460-250G
BT130460-1KGEA 121.2 USD InStock
BT130460-1KG BT130460-250G
Benzotrifluoride ≥99%
Benzotrifluoride
CAS: 98-08-8; EC No: 202-635-0; MDL No: MFCD00000372; RTECS: XT9450000 UN No: UN2338; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C7H5F3; MW: 146.11 Melting Point: -29°; Boiling Point: 102°; Flash point: 12°C (54°F) Density (g/mL): 1.19; Refractive Index: 1.414

  • RTECS: XT9450000
  • Refractive index: 1.414
  • Liquid


Formula: C₇H₅F₃
Boiling Pt: 102 °C
Melting Pt: –29 °C
Density: 1.19 g/mL
Flash Pt: 12 °C (54 °F)
MDL Number: MFCD00000372
CAS Number: 98-08-8
EINECS: 202-635-0
UN: 2338
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05485864abd8b35273efdbb2e08a5ed79b907869&vid=05485864abd8b35273efdbb2e08a5ed79b907869&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty