Lead(II) chloride ≥99.999%
Supplier: BEANTOWN CHEMICAL - BTC
132150-100G
132150-25G
BT132150-100GEA
367
USD
BT132150-100G
BT132150-25G
Lead(II) chloride ≥99.999%
Lead(II) chloride
CAS: 7758-95-4; EC No: 231-845-5; MDL No: MFCD00011157; RTECS: OF9450000
UN No: UN2291; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: PbCl2; MW: 278.10
Melting Point: 501°; Boiling Point: 950°
Density (g/mL): 5.85
- RTECS: OF9450000
- Crystalline
Formula:
Cl₂Pb MW: 278.11 g/mol Boiling Pt: 950 Melting Pt: 496 Density: 5.85 g/mL |
MDL Number:
MFCD00011157 CAS Number: 7758-95-4 EINECS: 231-845-5 UN: 2291 ADR: 6.1 III Merck Index: 13,05422 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...