133500-10G
133500-50G
BT133500-10GEA
80
USD
InStock
BT133500-10G
BT133500-50G
Potassium selenocyanate ≥98.5%
Potassium selenocyanate
CAS: 3425-46-5; EC No: 222-320-1; MDL No: MFCD00011366; RTECS: VT4000000
UN No: UN3283; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: KSeCN; MW: 144.08
Melting Point: 147°
Density (g/mL): 2.35
Moisture Sensitive
- RTECS: VT4000000
- Moisture Sensitive
- Crystalline