138210-1G
138210-5G
BT138210-1GEA
160
USD
InStock
BT138210-1G
BT138210-5G
α-Bromo-4-chloro-2,6-difluorotoluene ≥97%
α-Bromo-4-chloro-2,6-difluorotoluene
CAS: 537013-52-8; MDL No: MFCD03094463
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C7H4BrClF2; MW: 241.46