1,3,5,7-Cyclooctatetraene ≥98% stabilised

Supplier: BEANTOWN CHEMICAL - BTC

140380-1G 140380-25G 140380-5G
BT140380-1GEA 52 USD
BT140380-1G BT140380-25G BT140380-5G
1,3,5,7-Cyclooctatetraene ≥98% stabilised
1,3,5,7-Cyclooctatetraene

CAS: 629-20-9; EC No: 211-080-3; MDL No: MFCD00004161 UN No: UN2358; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C8H8 ; MW: 104.15 Melting Point: -5°; Boiling Point: 142-143°; Flash point: 22°C (72°F) Density (g/mL): 0.925 ; Refractive Index: 1.537 Light Sensitive


  • Refractive index: 1.537
  • Light Sensitive
  • Liquid

Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 143
Melting Pt: -7
Density: 0.925 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00004161
CAS Number: 629-20-9
EINECS: 211-080-3
UN: 2358
ADR: 3 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR