Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fullerene, hydroxylated, C60(OH)n, powder

Supplier: BEANTOWN CHEMICAL - BTC

141030-100MG 141030-25MG
BT141030-100MGEA 560 USD InStock
BT141030-100MG BT141030-25MG
Fullerene, hydroxylated, C60(OH)n, powder
Fullerene, hydroxylated, C60(OH)n
MDL No: MFCD06202029 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: C; MW: 12.011


MDL Number: MFCD06202029
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty