Lutetium ≥99.9% (trace rare earth metals basis), pieces, distilled dendritic

Supplier: BEANTOWN CHEMICAL - BTC

141090-1G 141090-5G
BT141090-1GEA 223 USD
BT141090-1G BT141090-5G
Lutetium ≥99.9% (trace rare earth metals basis), pieces, distilled dendritic
Lutetium

CAS: 7439-94-3; EC No: 231-103-0; MDL No: MFCD00011098 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Lu; MW: 174.97 Melting Point: 1663°; Boiling Point: 3402° Density (g/mL): 9.84


  • Solid

Formula: Lu
MW: 174.97 g/mol
Boiling Pt: 3000
Melting Pt: 1663 °C
Density: 9.84 g/mL
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
ADR: 4.1 II
Merck Index: 12,05642

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR