Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vanadium(IV) oxide ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

143105-10G 143105-50G
BT143105-10GEA 76.28 USD InStock
BT143105-10G BT143105-50G
Vanadium(IV) oxide ≥99%
Vanadium(IV) oxide
CAS: 12036-21-4; EC No: 234-841-1; MDL No: MFCD00049702 UN No: UN3285; Haz Class: 6.1; Packing Group: II -100 Mesh Powder; Linear Formula: VO2; MW: 82.94 Melting Point: 1967° Density (g/mL): 4.339

  • -100 Mesh Powder


Formula: O₂V
Melting Pt: 1967 °C
Density: 4.339 g/mL
MDL Number: MFCD00049702
CAS Number: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty