Cerium(III) nitrate hydrate ≥99.995%

Supplier: BEANTOWN CHEMICAL - BTC

213310-100G 213310-25G
BT213310-100GEA 403 USD
BT213310-100G BT213310-25G
Cerium(III) nitrate hydrate ≥99.995%
Cerium(III) nitrate hydrate

EC No: 233-297-2; MDL No: MFCD00010926 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline; Molecular Formula: CeN3O9·xH2O; MW: 326.13 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Ce(NO₃)₃·xH₂O
MW: 326.13
Density: 1.9
MDL Number: MFCD00010926
CAS Number: 10108-73-3
EINECS: 233-297-2
ADR: 5.1 III
Merck Index: 12,02044

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR