217595-25G
217595-5G
BT217595-25GEA
211
USD
InStock
BT217595-25G
BT217595-5G
5-Hexen-1-ol ≥98%
5-Hexen-1-ol
CAS: 821-41-0; EC No: 212-477-4; MDL No: MFCD00002981
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: HOCH2(CH2)3CH=CH2; Molecular Formula: C6H12O; MW: 100.16
Boiling Point: 78-80°/25 mmHg; Flash point: 47°C (117°F)
Density (g/mL): 0.834; Refractive Index: 1.435
- Refractive index: 1.435
- Liquid