H66600-03
H66600-MD
AAH66600-03EA
364
USD
InStock
AAH66600-03
AAH66600-MD
(S)-2-Amino-3-cyclopentylpropanoic acid 95%
(S)-2-Amino-3-cyclopentylpropanoic acid
3-Cyclopentyl-L-alanine, 95%
Purity: 95%
CAS No.: 99295-82-6
Molecular Formula: C8H15NO2
Formula Weight: 157.21
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 10659781
MDL No.: MFCD01632069