H66803-03
H66803-06
H66803-MD
AAH66803-03EA
74.9
USD
InStock
AAH66803-03
AAH66803-06
AAH66803-MD
Fmoc-D-Lys(Mtt)-OH 98%
Fmoc-D-Lys(Mtt)-OH
N^a-Fmoc-N^e-(4-methyltrityl)-D-lysine, 98%
Purity: 98%
CAS No.: 198544-94-4
Molecular Formula: C41H40N2O4
Formula Weight: 624.78
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00270631