120-12-5UG
120-12-25UG
120-12-100UG
120-12-250UG
120-12-500UG
120-12-1MG
10772-480EA
132.81
USD
InStock
10772-480
10772-482
10772-484
10772-486
10772-488
10772-490
Human Recombinant Myostatin-Propeptide (from E. coli)
Proteins and Peptides
Mature Myostatin is obtained by proteolytic processing of a biologically-inactive precursor protein, which contains an N-terminal propeptide of 243 amino acid residues. Myostatin-Propeptide exhibits high binding affinity for myostatin, and has been shown to be a potent inhibitor of myostatin. Over-expression of myostatin-propeptide in mice resulted in large increases (up to 200%) in skeletal muscle mass, similar to those observed in myostatin knockout mice. Recombinant Human Myostatin-Propeptide is a 27.8 kDa protein consisting of 244 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.