Human Recombinant Follistatin (from E. coli), PeproTech
Supplier: PEPROTECH INC.
120-13-5UG 120-13-20UG 120-13-100UG 120-13-250UG 120-13-500UG 120-13-1MG
10772-492EA 132.81 USD InStock
10772-492 10772-494 10772-496 10772-498 10772-500 10772-502
Human Recombinant Follistatin (from E. coli), PeproTech
Proteins and Peptides
Follistatin is a secreted protein that binds to ligands of the TGF-β family and regulates their activity by inhibiting their access to signaling receptors. It was originally discovered as an activin antagonist whose activity suppresses expression and secretion of the pituitary hormone FSH (follicle stimulating hormone). In addition to being a natural antagonist, follistatin can inhibit the activity of other TGF-β ligands including BMP-2,-4,-6,-7, Myostatin, GDF-11, and TGF-β1. Follistatin is expressed in the pituitary, ovaries, decidual cells of the endometrium, and in some other tissues. Recombinant Human Follistatin is a 31.5 kDa protein containing 288 amino acids. Its primary structure contains three cysteine-rich domains (called FS domains), each followed by a protease-inhibitory kazal domain.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty