Human Recombinant Relaxin-3 (from E. coli)
Supplier: PEPROTECH INC.
130-10-5UG 130-10-25UG 130-10-100UG 130-10-250UG 130-10-500UG 130-10-1MG
10772-740EA 133.07 USD InStock
10772-740 10772-742 10772-744 10772-746 10772-748 10772-750
Human Recombinant Relaxin-3 (from E. coli)
Proteins and Peptides
Relaxin-3 is a secreted protein structurally related to insulin that is expressed primarily in the brain and central nervous system. Relaxin-3 has been identified as the ligand for the GPCR135 receptor, previously known as “somatostatin-like” or “angiotensin-like” peptide receptor, and has also been identified for binding specifically to the LGR7 receptor, previously identified as an “orphan” G protein-coupled receptor. Signaling by relaxin-3 through its target receptors is, most likely, part of a CNS processing system, activated in response to signaling by neuropeptides and other factors. Intracerebroventricular injections of relaxin-3 have been shown to cause a significant increase of food intake and body weight in Wistar rats. Recombinant Human Relaxin-3 is a 5.5 kDa, disulfide-linked, heterodimeric protein consisting of a 24 amino acid A-chain and a 27 amino acid B-chain.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty