Human Recombinant IL-11 (from E. coli)
Supplier: PEPROTECH INC.
AF-200-11-250UG AF-200-11-500UG AF-200-11-1MG
10778-314EA 3226.99 USD InStock
10778-314 10778-316 10778-318
Human Recombinant IL-11 (from E. coli)
Proteins and Peptides
IL-11 is a multifunctional cytokine produced by stromal cells, such as fibroblasts, epithelial cells and osteoclasts. It is expressed in a wide variety of tissues, including thymus, lung, bone, connective tissue and central nervous system. IL-11 plays an important regulatory role in hematopoiesis by stimulating growth of myeloid, erythroid and megakaryocyte progenitor cells. It also regulates bone metabolism, inhibits production of proinflammatory cytokines, and protects against gastromucosal injury. Recombinant Human IL-11 is a 19.3 kDa protein consisting of 179 amino acid residues.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty