Human Recombinant Oncostatin (from E. coli)
Supplier: PEPROTECH INC.
AF-300-10T-2UG AF-300-10T-10UG AF-300-10T-100UG AF-300-10T-250UG AF-300-10T-500UG AF-300-10T-1MG
10778-638EA 183.06 USD InStock
10778-638 10778-640 10778-642 10778-644 10778-646 10778-648
Human Recombinant Oncostatin (from E. coli)
Proteins and Peptides
Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi’s sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi’s sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF. Human OSM is active on murine cells. The human OSM gene encodes for a 252 amino acid polypeptide, containing 25 amino acid signal sequence for secretion and a 227 precursor protein. Proteolytic processing of this precursor removes an 18 amino acid C-terminal peptide and generates the mature OSM form. Recombinant Human Oncostatin M is a 23.6 kDa protein, containing 209 amino acid residues.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty