Human Recombinant IL-36RA (from E. coli)
Supplier: PEPROTECH INC.
200-36RA-5UG 200-36RA-25UG 200-36RA-100UG 200-36RA-250UG 200-36RA-500UG 200-36RA-1MG
10779-904EA 143.88 USD InStock
10779-904 10779-906 10779-908 10779-910 10779-912 10779-914
Human Recombinant IL-36RA (from E. coli)
Proteins and Peptides
The IL-1 family is comprised of 11 structurally related ligands, including the recently re-named IL-36RA (IL-1F5), IL-36α (IL-1F6), IL-36β (IL-1F8), and IL-36γ (IL-1F9). The interaction of IL-36 ligands with the IL-1Rrp2 receptor (IL-1R6) can induce various activities, including dendritic cell maturation and activation. IL-36RA can antagonize the NF-κB signaling induced by either IL-36α, β or γ by binding to the IL-1Rrp2 receptor in a manner that prevents the initiation of functional signaling. Recombinant Human IL-36RA is an
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty