210-15-2UG
210-15-10UG
210-15-50UG
210-15-100UG
210-15-250UG
210-15-500UG
210-15-1MG
10779-976EA
139.52
USD
InStock
10779-976
10779-978
10779-980
10779-982
10779-984
10779-986
10779-988
Mouse Recombinant IL-15 (from E. coli)
Proteins and Peptides
IL-15 is an immunomodulating cytokine that stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. IL-15 exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. Increased expression of IL-15 has been linked to rheumatoid arthritis, inflammatory bowel disease, and diseases affiliated with retroviruses HIV and HTLV-I. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells. Recombinant Murine IL-15 is a 13.3 kDa protein consisting of 115 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.