Human Recombinant TGF-alpha (from E. coli)
Supplier: PEPROTECH INC.
AF-100-16A-20UG AF-100-16A-100UG AF-100-16A-250UG AF-100-16A-500UG AF-100-16A-1MG
10781-698EA 183.06 USD InStock
10781-698 10781-700 10781-702 10781-704 10781-706
Human Recombinant TGF-alpha (from E. coli)
Proteins and Peptides
TGF-α is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-α induces anchorage-independent transformation in cultured cells. Human, murine and rat TGF-α are cross-species reactive. Recombinant Human TGF-α is a 50 amino acid polypeptide (5.5 kDa), which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty