Acetonitrile ACS

Supplier: EPREDIA
Danger

10021 10020
100503-628EA 275.24 USD
100503-628 100503-626
Acetonitrile ACS
Acetonitrile

A non-carcinogenic substitute for Propylene Oxide, used in tissueprocessing for TEM. Good for infiltration with EMbed-812 or other Epon-812 substitutes. Tarnowski, sB.I. et al, EMSA proceedings, Detroit, MI page 38 (1984).

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.7822 g/cm³ (6.527 lbs/gal.)
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3 II
Merck Index: 13,00071
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c64e994b81e0fd2ff1018db8a5aeafff4e4ded16&vid=c64e994b81e0fd2ff1018db8a5aeafff4e4ded16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR