Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL23R Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 753317]

Supplier: R&D Systems
FAB16861G-025 FAB16861G-100
102880-926EA 258.86 USD InStock
102880-926 102881-272
Anti-IL23R Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 753317]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 753317
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty