Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SLAMF6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 292811]

Supplier: R&D Systems
FAB19081A-025 FAB19081A-100
102881-574EA 201.02 USD InStock
102881-574 102881-576
Anti-SLAMF6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 292811]
Antibodies
Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: 292811
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty