H61623-03
H61623-06
AAH61623-03EA
37.7
USD
InStock
AAH61623-03
AAH61623-06
4-Bromo-2,6-difluorobenzyl alcohol 97%
4-Bromo-2,6-difluorobenzyl alcohol
Purity: 97%
CAS No.: 162744-59-4
Molecular Formula: C7H5BrF2O
Formula Weight: 223.01
Physical Form: Solid
Melting Point: 76-81°
MDL No.: MFCD03094461