Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD107b (LAMP-2) Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H4B4]

Supplier: Biolegend
354305 354306
354305-BLEA 120.5 USD InStock
354305-BL 354306-BL
Anti-CD107b (LAMP-2) Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H4B4]
Antibodies
FITC anti-human CD107b (LAMP-2) [H4B4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107b (LAMP-2)
Clonality: Monoclonal
Clone: H4B4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty