Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Acetylated Lysine Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 15G10]

Supplier: Biolegend
623407 623408
10019-836EA 198.1 USD InStock
10019-836 10019-838
Anti-Acetylated Lysine Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 15G10]
Antibodies
PE/Cy7 anti-Acetylated Lysine [15G10]; Isotype: Mouse IgG2b, κ; Reactivity: Acetylated lysine residues; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Acetylated Lysine
Clonality: Monoclonal
Clone: 15G10
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Acetylated lysine residues

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty