Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD22 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S-HCL-1]

Supplier: Biolegend
363505 363506
10019-530EA 140.7 USD InStock
10019-530 10019-532
Anti-CD22 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S-HCL-1]
Antibodies
APC anti-human CD22 [S-HCL-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: S-HCL-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty