Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dichloropyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

072918-250MG 072918-1G 072918-5G
102527-436EA 387.69 USD InStock
102527-436 102527-438 102527-440
4,5-Dichloropyrimidine ≥95%
4,5-Dichloropyrimidine
Hazard: Irritant/Refrigerate


Formula: C₄H₂Cl₂N₂
Boiling Pt: 55…56 °C (5 mmHg)
MDL Number: MFCD09743880
CAS Number: 6554-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty