Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Isopropylcyclohexanecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

073515-10G 073515-100G 073515-500G
102530-576EA 23.08 USD InStock
102530-576 102530-578 102530-580
trans-4-Isopropylcyclohexanecarboxylic acid ≥95%
trans-4-Isopropylcyclohexanecarboxylic acid
Hazard: Irritant


Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Melting Pt: 94 °C
MDL Number: MFCD04004145
CAS Number: 7077-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty