Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Naphthyl)phenylboronic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

075164-1G 075164-5G 075164-10G
102540-084EA 107.69 USD InStock
102540-084 102540-086 102540-088
4-(1-Naphthyl)phenylboronic acid ≥95%
4-(1-Naphthyl)phenylboronic acid
Hazard: Irritant


Formula: C₁₆H₁₃BO₂
MDL Number: MFCD08669639
CAS Number: 870774-25-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty