Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥95%

Supplier: MATRIX SCIENTIFIC MS

075540-1G 075540-5G 075540-10G
102541-818EA 61.54 USD
102541-818 102541-820 102541-822
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥95%
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate

Hazard: Irritant

Formula: C₁₂H₂₁NO₈
MW: 307.30 g/mol
MDL Number: MFCD03093639
CAS Number: 163343-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR