3,5-Dimethoxyaniline ≥95%
Supplier: MATRIX SCIENTIFIC MS
075744-25G
075744-100G
102542-818EA
136.92
USD
102542-818
102542-820
3,5-Dimethoxyaniline ≥95%
3,5-Dimethoxyaniline
Hazard: Toxic
Formula:
C₈H₁₁NO₂ MW: 153.18 g/mol Boiling Pt: 303 Melting Pt: 53…57 °C |
MDL Number:
MFCD00008392 CAS Number: 10272-07-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...