Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nilotinib ≥95%

Supplier: MATRIX SCIENTIFIC MS

075955-250MG 075955-1G
102543-794EA 498.46 USD InStock
102543-794 102543-796
Nilotinib ≥95%
Nilotinib
Hazard: Irritant


Formula: C₂₈H₂₂F₃N₇O
MDL Number: MFCD09833716
CAS Number: 641571-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty