Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-2-urea ≥95%

Supplier: MATRIX SCIENTIFIC MS

075990-10G 075990-100G
102543-978EA 36.92 USD InStock
102543-978 102543-980
1-Phenyl-2-urea ≥95%
1-Phenyl-2-urea
Hazard: Irritant


Formula: C₇H₈N₂O
MDL Number: MFCD00007944
CAS Number: 64-10-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty