Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine ≥95%

Supplier: MATRIX SCIENTIFIC MS

076503-250MG 076503-1G
102546-542EA 924.62 USD InStock
102546-542 102546-544
1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine ≥95%
1-(3-Chlorophenyl)-2,2,2-trifluoroethylamine
Hazard: Irritant


Formula: C₈H₇ClF₃N
MDL Number: MFCD07374613
CAS Number: 886368-66-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty