Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

077039-500MG 077039-1G
102664-286EA 995.38 USD InStock
102664-286 102664-288
(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid ≥97%
(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid
Hazard: Irritant


Formula: C₁₇H₂₄O₅
MDL Number: MFCD11112069
CAS Number: 387868-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty