Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-1-carbaldehyde

Supplier: MATRIX SCIENTIFIC MS

077288-500MG 077288-1G 077288-5G
102665-424EA 516.92 USD InStock
102665-424 102665-426 102665-428
3-Methyl-4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-1-carbaldehyde
3-Methyl-4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-1-carbaldehyde
Hazard: Irritant


Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD01099157
CAS Number: 104310-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty