Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Oxatricyclo[4.3.1.1~3,8~]undecan-5-one

Supplier: MATRIX SCIENTIFIC MS

077625-500MG 077625-1G 077625-5G
102667-172EA 304.62 USD InStock
102667-172 102667-174 102667-176
4-Oxatricyclo[4.3.1.1~3,8~]undecan-5-one
4-Oxatricyclo[4.3.1.1~3,8~]undecan-5-one
Hazard: Irritant


Formula: C₁₀H₁₄O₂
MDL Number: MFCD00184584
CAS Number: 21898-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty