Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyanobenzoyl chloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

083103-1G 083103-10G
102838-924EA 100 USD InStock
102838-924 102838-926
3-Cyanobenzoyl chloride ≥95%
3-Cyanobenzoyl chloride
Hazard: Corrosive


Formula: C₈H₄ClNO
MW: 165.58
Boiling Pt: 125…129 °C (11 mmHg)
Melting Pt: 42…44 °C
Flash Pt: 113 °C
MDL Number: MFCD00011545
CAS Number: 1711-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty