2-(3-Fluorobenzoyl)oxazole ≥97%
Supplier: MATRIX SCIENTIFIC MS
086622-1G
086622-2G
086622-5G
102849-646EA
795.38
USD
102849-646
102849-648
102849-650
2-(3-Fluorobenzoyl)oxazole ≥97%
2-(3-Fluorobenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₀H₆FNO₂ MW: 191.16 g/mol |
MDL Number:
MFCD07699346 CAS Number: 898759-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...